(3,4-Dimethoxyphenyl)acetonitrile ≥98%

Synonyms: 3,4-Dimethoxyphenylacetonitrile, Homoveratronitrile, 3,4-Dimethoxybenzyl cyanide

126349-100G 126349-500G
220035-361EA 128.15 USD
220035-361 220035-362
(3,4-Dimethoxyphenyl)acetonitrile ≥98%
(3,4-Dimethoxyphenyl)acetonitrile
Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 60 - 62 °C
MDL Number: MFCD00001911
CAS Number: 93-17-4
EINECS: 202-225-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR