3,5-Dimethoxybenzaldehyde ≥98%

126292-10G 126292-50G
220035-352EA 180.92 USD
220035-352 220035-353
3,5-Dimethoxybenzaldehyde ≥98%
3,5-Dimethoxybenzaldehyde
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 277 °C (1013 hPa)
Melting Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00003366
CAS Number: 7311-34-4
EINECS: 230-772-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR