1,2-Difluorobenzene ≥98%

126152-10G 126152-50G
220034-793EA 74.09 USD
220034-793 220034-794
1,2-Difluorobenzene ≥98%
1,2-Difluorobenzene
Formula: C₆H₄F₂
MW: 114.09 g/mol
Boiling Pt: 92 °C (1013 hPa)
Melting Pt: -34 °C
Density: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000284
CAS Number: 367-11-3
EINECS: 206-680-7
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR