2,4-Dichloro-6-nitroaniline ≥98%

Danger

126004-5G 126004-25G
220035-332EA 2107.69 USD
220035-332 220035-331
2,4-Dichloro-6-nitroaniline ≥98%
2,4-Dichloro-6-nitroaniline
Formula: C₆H₄Cl₂N₂O₂
MW: 207.02 g/mol
Melting Pt: 110 - 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00007664
CAS Number: 2683-43-4
EINECS: 220-241-7
UN: 2237
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR