α,α'-Dibromo-m-xylene ≥97%

Warning

Synonyms: 1,3-Bis(bromomethyl)benzene, m-Xylylene dibromide

125911-10G 125911-25G
220035-324EA 193.85 USD
220035-324 220035-325
α,α'-Dibromo-m-xylene ≥97%
α,α'-Dibromo-m-xylene
Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 135…140 °C (20 mmHg)
Melting Pt: 76…78 °C
Density: 1.959 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000178
CAS Number: 626-15-3
EINECS: 210-931-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR