2,5-Dibromoaniline ≥98%

125881-5G 125881-25G
220035-321EA 43.83 USD
220035-321 220035-320
2,5-Dibromoaniline ≥98%
2,5-Dibromoaniline
Formula: C₆H₅Br₂N
Boiling Pt: 281 °C (1013 hPa)
Melting Pt: 54.5 - 55 °C
MDL Number: MFCD00007636
CAS Number: 3638-73-1
EINECS: 222-865-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR