2,3-Diaminopyridine

Warning

Synonyms: Pyridine-2,3-diyldiamine

125857-5G 125857-25G 125857-1G
220035-319EA 154 USD
220035-319 220035-318 220035-317
2,3-Diaminopyridine
2,3-Diaminopyridine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 148…150 °C (5 mmHg)
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00006319
CAS Number: 452-58-4
EINECS: 207-200-9
UN: 2671
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR