Citraconic anhydride

Synonyms: 3-Methylfuran-2,5-dione

125318-100G 125318-25G 125318-500G
220035-294EA 138.62 USD
220035-294 220035-295 220035-296
Citraconic anhydride
Citraconic anhydride
Formula: C₅H₄O₃
MW: 112.08 g/mol
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: 7…8 °C
Density: 1.243 g/cm³ (20 °C)
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005522
CAS Number: 616-02-4
EINECS: 210-459-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR