4-Chlorothiophenol

Warning

Synonyms: 1-Chloro-4-mercaptobenzene, 4-Chlorobenzene-1-thiol, 4-Chlorobenzenethiol, p-Chlorothiophenol

125237-5G 125237-500G 125237-100G
220035-291EA 47.38 USD
220035-291 220035-290 220035-289
4-Chlorothiophenol
4-Chlorothiophenol
Formula: C₆H₅ClS
MW: 144.62 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 49…53 °C
Density: 1.224 g/cm³ (60 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004847
CAS Number: 106-54-7
EINECS: 203-408-9
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR