4-Formylbenzoic acid ≥97%

Synonyms: Terephthalaldehydic acid, 4-Carboxybenzaldehyde

124915-10G 124915-25G 124915-100G
220035-259EA 64.08 USD
220035-259 220035-260 220035-258
4-Formylbenzoic acid ≥97%
4-Formylbenzoic acid
Formula: C₈H₆O₃
MW: 150.13 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 148 °C
MDL Number: MFCD00006951
CAS Number: 619-66-9
EINECS: 210-607-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR