Undec-10-enoic acid ≥98%

Warning

Synonyms: Undecylenic-10 acid, Undecenoic acid, 10-Undecenoic acid, Undecylenic acid, Undecenylic acid

124672-100ML 124672-500ML
220035-233EA 29.83 USD
220035-233 220035-234
Undec-10-enoic acid ≥98%
Undec-10-enoic acid
Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 24 °C
Density: 0.91 g/cm³ (20 °C)
Flash Pt: 146 °C
MDL Number: MFCD00004442
CAS Number: 112-38-9
EINECS: 203-965-8
Merck Index: 12,09983

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR