tert-Butyl bromoacetate ≥98%

Synonyms: Bromoacetic tert-butyl ester

124230-10G 124230-250G 124230-50G
220034-862EA 33.82 USD
220034-862 220034-863 220034-864
tert-Butyl bromoacetate ≥98%
tert-Butyl bromoacetate
Formula: BrCH₂COOC(CH₃)₂
MW: 195.06 g/mol
Boiling Pt: 50 °C (1013 hPa)
Density: 1.338 g/cm³ (20 °C)
MDL Number: MFCD00000188
CAS Number: 5292-43-3
EINECS: 226-133-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR