3-Bromoquinoline ≥98%

Warning

124133-100G 124133-25G
220034-980EA 330.62 USD
220034-980 220034-981
3-Bromoquinoline ≥98%
3-Bromoquinoline
Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 274 °C (1013 hPa)
Melting Pt: 15 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006767
CAS Number: 5332-24-1
EINECS: 226-237-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR