1-Bromo-3-methylbutane ≥96%

Danger

Synonyms: 3-Methylbutylbromide

124095-100G 124095-5G 124095-500G
220034-975EA 48.46 USD
220034-975 220034-977 220034-976
1-Bromo-3-methylbutane ≥96%
1-Bromo-3-methylbutane
Formula: C₅H₁₁Br
MW: 151.05 g/mol
Boiling Pt: 119 °C (1013 hPa)
Melting Pt: -112 °C
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000253
CAS Number: 107-82-4
EINECS: 203-522-9
UN: 2341
ADR: 3,III
Merck Index: 13,05130

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR