1-Bromo-3-chlorobenzene ≥99%

Warning

Synonyms: 3-Bromochlorobenzene

124036-25G 124036-100G
220034-967EA 63.75 USD
220034-967 220035-225
1-Bromo-3-chlorobenzene ≥99%
1-Bromo-3-chlorobenzene
Formula: C₆H₄BrCl
MW: 191.45 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: –21.2 °C
Density: 1.63 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000568
CAS Number: 108-37-2
EINECS: 203-575-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR