Bromoacetaldehyde diethyl acetal ≥97%

Synonyms: 2-Bromo-1,1-diethoxyethane

123986-500G 123986-100G 123986-1KG
220035-224EA 187.38 USD
220035-224 220035-222 220035-223
Bromoacetaldehyde diethyl acetal ≥97%
Bromoacetaldehyde diethyl acetal
Formula: BrH₂CCH(OCH₂CH₃)₂
MW: 197.07 g/mol
Boiling Pt: 66 °C (1013 hPa)
Density: 1.27 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000214
CAS Number: 2032-35-1
EINECS: 217-989-1
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR