α,α'-Dichloro-o-xylene ≥98%

Synonyms: o-Xylylene Dichloride, o-Xylylene chloride, α,α′-Dichloro-o-xylene, α,α'-Dichloro-o-xylene, 1,2-Bis(chloromethyl)benzene, Alpha,Alpha’-Dichloro-o-xylene, 1,2-Bis(chloromethyl)benzene, o-Xylene dichloride

123889-100G 123889-25G
220035-177EA 178.77 USD
220035-177 220035-178
α,α'-Dichloro-o-xylene ≥98%
α,α'-Dichloro-o-xylene
Formula: C₈H₈Cl₂
MW: 175.06 g/mol
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: 54…55 °C
Density: 1.393 g/cm³ (0 °C)
MDL Number: MFCD00000903
CAS Number: 612-12-4
EINECS: 210-291-8
UN: 2928
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR