4-(Benzyloxy)benzaldehyde ≥97%

123714-25G 123714-100G
220035-174EA 91.43 USD
220035-174 220034-785
4-(Benzyloxy)benzaldehyde ≥97%
4-(Benzyloxy)benzaldehyde
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 366 °C (1013 hPa)
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00003387
CAS Number: 4397-53-9
EINECS: 224-527-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR