5-Aminopentan-1-ol ≥95%

Synonyms: 5-Amino-1-pentanol

123048-50G 123048-10G
220035-044EA 188.46 USD
220035-044 220034-397
5-Aminopentan-1-ol ≥95%
5-Aminopentan-1-ol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 36 °C
Density: 0.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008237
CAS Number: 2508-29-4
EINECS: 219-718-2
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR