2-Aminobenzyl alcohol ≥98%

Synonyms: (2-Aminophenyl)methanol, 2-Hydroxymethylaniline, 2-(Hydroxymethyl)aniline, 2-Aminobenzyl Alcohol

122831-25G 122831-10G
220034-385EA 70.86 USD
220034-385 220034-384
2-Aminobenzyl alcohol ≥98%
2-Aminobenzyl alcohol
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007749
CAS Number: 5344-90-1
EINECS: 226-293-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR