Alizarine

Warning

Synonyms: Alizarin, 1,2-Dihydroxy-9,10-anthraquinone, 1,2-Dihydroxy-9,10-anthracenedione, 1,2-Dihydroxyanthraquinone

122777-500G 122777-100G
220034-379EA 961.8 USD
220034-379 220034-966
Alizarine
Alizarine
Formula: C₁₄H₈O₄
MW: 240.22 g/mol
Boiling Pt: 430 °C (1013 hPa)
Melting Pt: 290 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001201
CAS Number: 72-48-0
EINECS: 200-782-5
Merck Index: 13,00246
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4380c49ce346457267379c6e7651572087bdd9d2&vid=d9c7b41b4b279049d0b0637d6806082e72600025&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR