(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol

Synonyms: (2,2-Dimethyl-1,3-dioxolan-4-yl)methanol, 2,2-Dimethyl-1,3-dioxolan-4-ylmethanol, DL-1,2-Isopropylidenglycerol, 1,2-O-Isopropylideneglycerol, Solketal, (±)-1,2-O-Isopropylidenglycerol, DL-1,2-Isopropylideneglycerol

122696-3KG 122696-500G 122696-100G
220034-900EA 356.46 USD
220034-900 220034-901 220035-043
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 188 °C (1013 hPa)
Density: 1.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00063238
CAS Number: 100-79-8
EINECS: 202-888-7
Merck Index: 13,05233

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR