Ethyl levulinate ≥99%

Synonyms: Ethyl 4-oxopentanoate, 4-Oxovaleric acid ethyl ester, Ethyl 4-oxovalerate, 4-Ketovaleric acid ethyl ester, 4-Oxopentanoic acid ethyl ester, Levulinic acid ethyl ester

122629-100G 122629-500G
220035-036EA 63.54 USD
220035-036 220035-037
Ethyl levulinate ≥99%
Ethyl levulinate
Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 93 °C (1013 hPa)
Density: 1.01 g/cm³ (20 °C)
MDL Number: MFCD00009209
CAS Number: 539-88-8
EINECS: 208-728-2
Merck Index: 13,03852

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR