3-Hydroxy-2-pyridone

122505-25G 122505-100G
220034-859EA 193.85 USD
220034-859 220034-858
3-Hydroxy-2-pyridone
3-Hydroxy-2-pyridone
Formula: C₅H₅NO₂
MW: 111.1 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 244…246 °C
Storage Temperature: Ambient
MDL Number: MFCD00006271
CAS Number: 16867-04-2
EINECS: 240-887-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR