Neocuproine hydrate

Synonyms: 2,9-Dimethyl-1,10-phenanthroline hydrate

121908-25G 121908-5G
220034-820EA 274.62 USD
220034-820 220034-821
Neocuproine hydrate
Neocuproine hydrate
Formula: C₁₄H₁₂N₂·H₂O
MW: 226.28 g/mol
CAS Number: 654054-57-6
EINECS: 207-601-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR