2-Deoxy-D(-)-ribose

Synonyms: 2-Deoxy-D(-)-erythropentose, Thyminose, 2-Deoxy-D(-)-arabinose, 2'-Deoxy-D-Ribose, (3S,4R)-3,4,5-Trihydroxypentanal

121649-1G 121649-25G 121649-5G
220031-988EA 33.97 USD
220031-988 220031-989 220031-990
2-Deoxy-D(-)-ribose
2-Deoxy-D(-)-ribose
Formula: C₅H₁₀O₄
MW: 134.13 g/mol
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 95 - 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00135904
CAS Number: 533-67-5
EINECS: 208-573-0
Merck Index: 13,02925

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR