3,5-Dinitrobenzoic acid ≥99%

Danger

121258-100G 121258-500G
220034-765EA 48.68 USD
220034-765 220034-766
3,5-Dinitrobenzoic acid ≥99%
3,5-Dinitrobenzoic acid
Formula: C₇H₄N₂O₆
MW: 212.12 g/mol
Melting Pt: 204 - 208 °C
Density: 1.683 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007253
CAS Number: 99-34-3
EINECS: 202-751-1
UN: 1325
ADR: 4.1,III
Merck Index: 13,03304

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR