2,3,5-Triiodobenzoic acid ≥98%

Warning

Synonyms: TIBA

120979-100G 120979-10G
220034-374EA 425.38 USD
220034-374 220034-375
2,3,5-Triiodobenzoic acid ≥98%
2,3,5-Triiodobenzoic acid
Formula: C₇H₃I₃O₂
MW: 499.81 g/mol
Boiling Pt: 416 °C (1013 hPa)
Melting Pt: 225.9 °C
Storage Temperature: Freezer
MDL Number: MFCD00002420
CAS Number: 88-82-4
EINECS: 201-859-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR