2-Fluorobenzoyl chloride ≥99%

Danger

Synonyms: o-Fluorobenzoyl chloride

120847-100G 120847-25G
220034-358EA 84.11 USD
220034-358 220034-359
2-Fluorobenzoyl chloride ≥99%
2-Fluorobenzoyl chloride
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 206 °C (1013 hPa)
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000656
CAS Number: 393-52-2
EINECS: 206-887-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR