2-Hydroxy-m-anisaldehyde ≥99%

Synonyms: o-Vanillin, 2-Hydroxy-3-methoxybenzaldehyde

120804-100G 120804-10G 120804-500G
220034-354EA 108.77 USD
220034-354 220034-355 220034-356
2-Hydroxy-m-anisaldehyde ≥99%
2-Hydroxy-m-anisaldehyde
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 40…42 °C
Density: 1.2039 g/cm³ (45 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003322
CAS Number: 148-53-8
EINECS: 205-715-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR