4-Pyridone

Warning

Synonyms: 4-Hydroxypyridine, γ-Pyridone, 4-Pyridinol, Pyridin-4-ol, 1,4-Dihydro-4-Pyridinone

120618-500G 120618-100G 120618-25G
220034-728EA 754.92 USD
220034-728 220034-726 220034-727
4-Pyridone
4-Pyridone
Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 381 °C (1013 hPa)
Melting Pt: 146…148 °C
MDL Number: MFCD00006419
CAS Number: 626-64-2
EINECS: 210-958-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR