4-Fluorobenzoyl chloride ≥98%

Danger

Synonyms: p-Fluorobenzoyl chloride

119946-25G 119946-100G 119946-5G
220034-675EA 58.37 USD
220034-675 220034-674 220034-676
4-Fluorobenzoyl chloride ≥98%
4-Fluorobenzoyl chloride
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000684
CAS Number: 403-43-0
EINECS: 206-961-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR