2,6-Dichlorophenolindophenol sodium salt hydrate ACS, Sigma-Aldrich®

Synonyms: DPIP, DCIP

119814-5G 119814-10G
220034-221EA 55.15 USD
220034-221 220034-220
2,6-Dichlorophenolindophenol sodium salt hydrate ACS, Sigma-Aldrich®
2,6-Dichlorophenolindophenol sodium salt hydrate
Formula: C₁₂H₆Cl₂NNaO₂·xH₂O
MW: 290.08 g/mol (anhy)
Melting Pt: > 300 °C
MDL Number: MFCD00150014
CAS Number: 1266615-56-8
EINECS: 210-640-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR