2,4,6-Triisopropylbenzenesulfonyl chloride ≥97%

Danger

Synonyms: 2,4,6-Triisopropylbenzene-1-sulphonyl chloride

119490-25G 119490-100G
220033-988EA 82.92 USD
220033-988 220033-987
2,4,6-Triisopropylbenzenesulfonyl chloride ≥97%
2,4,6-Triisopropylbenzenesulphonyl chloride
Formula: C₁₅H₂₃ClO₂S
MW: 302.87 g/mol
Melting Pt: 93 - 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00007433
CAS Number: 6553-96-4
EINECS: 229-479-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR