3,3-Dimethyl-1-butene ≥95%

119059-250ML 119059-50ML
220034-207EA 239.08 USD
220034-207 220034-208
3,3-Dimethyl-1-butene ≥95%
3,3-Dimethyl-1-butene
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 41 °C (1013 hPa)
Melting Pt: –115 °C
Density: 0.6683 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00008853
CAS Number: 558-37-2
EINECS: 209-195-9
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR