1,5-Hexadien-3-ol

Synonyms: Hexa-1,5-dien-3-ol

118559-25G 118559-10G
220034-670EA 595.38 USD
220034-670 220034-669
1,5-Hexadien-3-ol
1,5-Hexadien-3-ol
Formula: C₆H₁₀O
MW: 98.14 g/mol
MDL Number: MFCD00008662
CAS Number: 924-41-4
EINECS: 213-102-7
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR