trans-Anethole ≥99%

Synonyms: (E)-1-Methoxy-4-(prop-1-en-1-yl)benzene

117870-25ML 117870-100ML
220034-655EA 72.37 USD
220034-655 220034-654
trans-Anethole ≥99%
trans-Anethole
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 236 °C (1013 hPa)
Melting Pt: 21…23 °C
Density: 0.9883 g/cm³ (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD00009284
CAS Number: 4180-23-8
EINECS: 224-052-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR