Phenylacetylene ≥98%

117706-100ML 117706-25ML
220033-664EA 168 USD
220033-664 220033-665
Phenylacetylene ≥98%
Phenylacetylene
Formula: C₈H₆
MW: 102.14 g/mol
Boiling Pt: 144 °C (1013 hPa)
Melting Pt: -44.8 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008570
CAS Number: 536-74-3
EINECS: 208-645-1
UN: 3295
ADR: 3,III
Merck Index: 13,03892

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR