Sodium barbiturate

11715-100G 11715-500G
220034-601EA 158.31 USD
220034-601 220034-602
Sodium barbiturate
Sodium barbiturate
Formula: C₄H₃N₂NaO₃
Storage Temperature: Ambient
CAS Number: 4390-16-3
EINECS: 224-507-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR