2-Methyl-1,3-cyclopentanedione ≥99%

Synonyms: 2-Methylcyclopentane-1,3-dione

117021-25G 117021-5G
220034-597EA 57.94 USD
220034-597 220034-598
2-Methyl-1,3-cyclopentanedione ≥99%
2-Methyl-1,3-cyclopentanedione
Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 213 °C (1013 hPa)
Density: 1.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001406
CAS Number: 765-69-5
EINECS: 212-153-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR