(±)-3-Phenylbutyric acid ≥98%

Synonyms: (±)-3-Phenylbutanoic acid

116807-25G 116807-5G
220033-892EA 359.69 USD
220033-892 220033-893
(±)-3-Phenylbutyric acid ≥98%
(±)-3-Phenylbutyric acid
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00002725
CAS Number: 4593-90-2
EINECS: 224-987-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR