2-Chloro-6-methylpyridine

Warning

Synonyms: 6-Chloro-2-picoline

116335-25G 116335-5G
220033-882EA 96 USD
220033-882 220033-883
2-Chloro-6-methylpyridine
2-Chloro-6-methylpyridine
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 64 °C (1013 hPa)
Density: 1.1506 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006245
CAS Number: 18368-63-3
EINECS: 242-241-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR