2-Chloro-4-methylpyridine

Warning

Synonyms: 2-Chloro-4-picoline

116327-25ML 116327-5ML
220033-880EA 273.54 USD
220033-880 220033-881
2-Chloro-4-methylpyridine
2-Chloro-4-methylpyridine
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 194…195 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.456 g/cm³ (25 °C)
MDL Number: MFCD00023418
CAS Number: 3678-62-4
EINECS: 222-951-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR