2-Chloro-3-hydroxypyridine

Warning

Synonyms: 2-Chloro-3-pyridinol

116203-5G 116203-25G
220034-583EA 74.42 USD
220034-583 220034-582
2-Chloro-3-hydroxypyridine
2-Chloro-3-hydroxypyridine
Formula: C₅H₄ClNO
MW: 129.55 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 170…171 °C
MDL Number: MFCD00006235
CAS Number: 6636-78-8
EINECS: 229-635-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR