2-Bromo-3-hydroxypyridine

Warning

Synonyms: 2-Bromo-3-pyridinol

116173-25G 116173-5G
220034-579EA 163.69 USD
220034-579 220034-580
2-Bromo-3-hydroxypyridine
2-Bromo-3-hydroxypyridine
Formula: C₅H₄BrNO
MW: 174 g/mol
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 185…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00006220
CAS Number: 6602-32-0
EINECS: 229-547-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR