2,5-Dibromo-p-xylene ≥98%

Synonyms: 2,5-Dibromo-1,4-dimethylbenzene, 1,4-Dibromo-2,5-dimethylbenzene

116157-100G 116157-25G
220034-577EA 182 USD
220034-577 220034-578
2,5-Dibromo-p-xylene ≥98%
2,5-Dibromo-p-xylene
Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 261 °C (1013 hPa)
Melting Pt: 75 °C
MDL Number: MFCD00000091
CAS Number: 1074-24-4
EINECS: 214-038-2
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR