(R)-(-)-2-Aminobutyric acid ≥99%

116122-1G 116122-5G
220034-572EA 36.62 USD
220034-572 220034-573
(R)-(-)-2-Aminobutyric acid ≥99%
(R)-(-)-2-Aminobutyric acid
Formula: C₄H₉NO₂
MDL Number: MFCD00064414
CAS Number: 2623-91-8
EINECS: 220-084-4

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR