2-Bromo-m-xylene ≥98%

Warning

Synonyms: 2-Bromo-1,3-dimethylbenzene, 2,6-Dimethylbromobenzene

115851-5G 115851-25G
220034-561EA 45.02 USD
220034-561 220034-560
2-Bromo-m-xylene ≥98%
2-Bromo-m-xylene
Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 206 °C (1013 hPa)
Melting Pt: –10 °C
Density: 1.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000075
CAS Number: 576-22-7
EINECS: 209-397-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR