3-Nitro-o-toluidine ≥97%

Danger

Synonyms: 3-Nitro-o-methylaniline, 3-Nitro-2-methylaniline, 2-Methyl-3-nitroaniline, 2-Amino-6-nitrotoluene

115843-100G 115843-25G
220034-557EA 162.62 USD
220034-557 220034-558
3-Nitro-o-toluidine ≥97%
3-Nitro-o-toluidine
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 87…89 °C
Density: 0.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007731
CAS Number: 603-83-8
EINECS: 210-059-6
UN: 2660
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR