ω-Bromoacetophenone ≥98%

Warning

Synonyms: α-Bromoacetophenone, Phenacyl bromide, 2-Bromoacetophenone

115835-500G 115835-5G 115835-100G
220034-554EA 198.15 USD
220034-554 220034-555 220034-553
ω-Bromoacetophenone ≥98%
ω-Bromoacetophenone
Formula: (C₆H₅)C(O)CH₂Br
MW: 199.05 g/mol
Boiling Pt: 253 °C (1013 hPa)
Melting Pt: 50 °C
Density: 1.647 g/cm³ (20 °C)
Flash Pt: ≥ 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000195
CAS Number: 70-11-1
EINECS: 200-724-9
UN: 2645
ADR: 6.1,II
Merck Index: 13,01386

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR